missing translation for 'onlineSavingsMsg'
Learn More

L-Valyl-L-phenylalanine, MedChemExpress

Product Code. 16485086
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485086 100 mg 100mg
16440477 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16485086 Supplier Medchem Express Supplier No. HY107378
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress L-Valyl-L-phenylalanine (Valylphenylalanine; H-VAL-PHE-OH) has been reported as biocompatible polymer.

L-Valyl-L-phenylalanine (Valylphenylalanine

Specifications

Chemical Name or Material L-Valyl-L-phenylalanine
Color White
CAS 3918-92-1
Physical Form Powder
Purity Grade Notes Research
Molecular Formula C14H20N2O3
Synonym Valylphenylalanine H-VAL-PHE-OH
Solubility Information DMSO : 100 mg/mL (378.33 mM; Need ultrasonic) ∣H2O : 66.67 mg/mL (252.23 mM; ultrasonic and adjust pH to 4 with HCl)
Quantity 100 mg
SMILES O=C([C@@H](N)C(C)C)N[C@H](C(O)=O)CC1=CC=CC=C1
Molecular Weight (g/mol) 264.32
Formula Weight 264.32
Percent Purity 98.95%
For Use With (Application) COVID-19-immunoregulation
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.