missing translation for 'onlineSavingsMsg'
Learn More

L-Valine, TRC

Product Code. 30133894
Change view
Click to view available options
Quantity:
10 mg
10 g
25 g
Unit Size:
10g
10mg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133894 25 g 25g
30133310 10 g 10g
30009921 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133894 Supplier Toronto Research Chemicals Supplier No. TRCV09420525G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-18-4
Molecular Formula C5 H11 N O2
Molecular Weight (g/mol) 117.15
Synonym L-Valine, Valine, L- (8CI), (+)-2-Amino-3-methylbutyric acid, (2S)-2-Amino-3-methylbutanoic acid, (2S)-2-Azaniumyl-3-methylbutanoate, (S)-2-Amino-3-methylbutanoic acid, (S)-2-Amino-3-methylbutyric acid, (S)-Valine, (S)-α-Amino-β-methylbutyric acid, 2-Amino-3-methylbutanoic acid, Butanoic acid, 2-amino-3-methyl-, (S)-, L-(+)-α-Aminoisovaleric acid, L-α-Amino-β-methylbutyric acid, NSC 76038, Valine
IUPAC Name (2S)-2-amino-3-methylbutanoic acid
SMILES CC(C)[C@H](N)C(=O)O

Specifications

InChI Formula InChI=1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)/t4-/m0/s1
Quantity 25 g
Formula Weight 117.079
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-Valine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.