missing translation for 'onlineSavingsMsg'
Learn More

L-Valine Methyl Ester Hydrochloride, TRC

Product Code. 30454643
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454643 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454643 Supplier Toronto Research Chemicals Supplier No. TRCV09432550G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Valine Methyl Ester Hydrochloride
CAS 6306-52-1
Molecular Formula C6 H13 N O2 . Cl H
Quantity 50 g
Synonym L-Valine','methyl ester','hydrochloride (1:1), L-Valine','methyl ester','hydrochloride (9CI), Valine','methyl ester','hydrochloride','L- (6CI,7CI,8CI), (S)-Valine methyl ester hydrochloride, Methyl (S)-2-amino-3-methylbutanoate hydrochloride, Methyl (S)-valinate hydrochloride, Methyl L-valinate hydrochloride, Methyl valinate hydrochloride, NSC 197198, NSC 22920, Valine methyl ester hydrochloride
InChI Formula InChI=1S/C6H13NO2.ClH/c1-4(2)5(7)6(8)9-3;/h4-5H,7H2,1-3H3;1H/t5-;/m0./s1
SMILES Cl.COC(=O)[C@@H](N)C(C)C
IUPAC Name methyl (2S)-2-amino-3-methylbutanoate;hydrochloride
Molecular Weight (g/mol) 167.63
Formula Weight 167.0713
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.