missing translation for 'onlineSavingsMsg'
Learn More

L-Valine Methyl Ester Hydrochloride, Mikromol™

Product Code. 17937954
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17937954 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17937954 Supplier Mikromol Supplier No. MM0487.060025
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

L-Valine Methyl Ester Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Valine Methyl Ester Hydrochloride
CAS 6306-52-1
Analyte or Component Names L-Valine Methyl Ester Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C6 H13 N O2 . Cl H
Certifications/Compliance ISO 17025
Synonym L-Valine, methyl ester, hydrochloride (1:1), L-Valine, methyl ester, hydrochloride (9CI), Valine, methyl ester, hydrochloride, L- (6CI,7CI,8CI), (S)-Valine methyl ester hydrochloride, Methyl (S)-2-amino-3-methylbutanoate hydrochloride, Methyl (S)-valinate hydrochloride, Methyl L-valinate hydrochloride, Methyl valinate hydrochloride, NSC 197198, NSC 22920, Valine methyl ester hydrochloride
InChI Formula InChI=1S/C6H13NO2.ClH/c1-4(2)5(7)6(8)9-3;/h4-5H,7H2,1-3H3;1H/t5-;/m0./s1
SMILES Cl.COC(=O)[C@@H](N)C(C)C
IUPAC Name methyl (2S)-2-amino-3-methylbutanoate;hydrochloride
Molecular Weight (g/mol) 167.63
Quantity 25 mg
Formula Weight 167.0713 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.