missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Valine methyl ester hydrochloride, 99%

Product Code. 10647172 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10647172 50 g Glass bottle 50g
10318320 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10647172 Supplier Thermo Scientific Chemicals Supplier No. 229260500
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6306-52-1 | C6H13ClNO2 | 166.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6306-52-1
Molecular Formula C6H13ClNO2
Molecular Weight (g/mol) 166.63
MDL Number MFCD00012497
InChI Key RFMIEHOYULVMNZ-UHFFFAOYNA-N
Synonym l-valine methyl ester hydrochloride, h-val-ome.hcl, methyl valinate hydrochloride, s-methyl 2-amino-3-methylbutanoate hydrochloride, h-l-val-ome hcl, methyl l-valinate hydrochloride, valine methyl ester hydrochloride, s-2-amino-3-methyl-butyric acid methyl ester hydrochloride, h-val-ome hcl, methyl 2s-2-amino-3-methylbutanoate hydrochloride
PubChem CID 111190
IUPAC Name methyl (2S)-2-amino-3-methylbutanoate;hydrochloride
SMILES [Cl].COC(=O)C(N)C(C)C

Specifications

Melting Point 171.0°C to 173.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (CH3)2CHCH(NH2)COOCH3·HCl
Quantity 50 g
Specific Rotation Condition +15.50° (24°C C=2,H2O)
Specific Rotation + 15.50
Solubility Information (5% in water) Clear colorless to light yellow
Formula Weight 167.64
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material L-Valine methyl ester hydrochloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.