missing translation for 'onlineSavingsMsg'
Learn More

L-Valine, MedChemExpress

Product Code. 16499126
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499126 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16499126 Supplier Medchem Express Supplier No. HYN0717
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-Valine (Valine) is a new nonlinear semiorganic material.

L-Valine (Valine) is a new nonlinear semiorganic material.

Specifications

Chemical Name or Material L-Valine
CAS 72-18-4
Color White
Purity Grade Notes Research
Molecular Formula C5H11NO2
Synonym Valine
Solubility Information H2O : 25 mg/mL (213.40 mM; Need ultrasonic)
SMILES N[C@@H](C(C)C)C(O)=O
Molecular Weight (g/mol) 117.15
Quantity 100 mg
Formula Weight 117.15
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.