missing translation for 'onlineSavingsMsg'
Learn More

L-Valine Benzyl Ester Hydrochloride, TRC

Product Code. 30450812
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30450812 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450812 Supplier Toronto Research Chemicals Supplier No. TRCV0942711G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Valine Benzyl Ester Hydrochloride
CAS 2462-34-2
Molecular Formula C12H18ClNO2
Quantity 1 g
Synonym L-Valine Phenylmethyl Ester Hydrochloride, Valine Benzyl Ester Hydrochloride, L-Valine Benzyl Ester Hydrochloride, (S)-Benzyl 2-Amino-3-methylbutanoate Hydrochloride, Benzyl L-Valinate Hydrochloride, L-Valine Benzyl Ester Hydrochloride
InChI Formula InChI=1S/C12H17NO2.ClH/c1-9(2)11(13)12(14)15-8-10-6-4-3-5-7-10;/h3-7,9,11H,8,13H2,1-2H3;1H/t11-;/m0./s1
SMILES Cl.CC(C)[C@H](N)C(=O)OCc1ccccc1
IUPAC Name benzyl (2S)-2-amino-3-methylbutanoate;hydrochloride
Molecular Weight (g/mol) 243.73
Formula Weight 243.1026
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.