missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Valine Amide, TRC
Specifications
Specifications
| Chemical Name or Material | L-Valine Amide |
| CAS | 3014-80-0 |
| Molecular Formula | C5H13ClN2O |
| Quantity | 100 g |
| Synonym | 2(S)- 2-Amino-3-methylbutanamide Monohydrochloride, (S)-2-Amino-3-methylbutanamide Monohydrochloride,, L-Monohydrochloride Valinamide, (2S)-2-Amino-3-methylbutanamide Hydrochloride, (S)-2-Amino-3-methylbutanamide Hydrochloride, (S)-Valinamide Hydrochloride, L-Valinamide Hydrochloride, L-Valine Amide Hydrochloride |
| InChI Formula | InChI=1S/C5H12N2O.ClH/c1-3(2)4(6)5(7)8;/h3-4H,6H2,1-2H3,(H2,7,8);1H/t4-;/m0./s1 |
| SMILES | Cl.CC(C)[C@H](N)C(=O)N |
| IUPAC Name | (2S)-2-amino-3-methylbutanamide;hydrochloride |
| Molecular Weight (g/mol) | 152.62 |
| Formula Weight | 152.0716 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?