missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Valine 99%
£29.40 - £742.00
Chemical Identifiers
CAS | 72-18-4 |
---|---|
Molecular Formula | C5H11NO2 |
Molecular Weight (g/mol) | 117.15 |
MDL Number | MFCD00064220 |
InChI Key | KZSNJWFQEVHDMF-BYPYZUCNSA-N |
Synonym | l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin |
PubChem CID | 6287 |
ChEBI | CHEBI:16414 |
IUPAC Name | (2S)-2-amino-3-methylbutanoic acid |
SMILES | CC(C)[C@H](N)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10473595
|
Thermo Scientific Chemicals
140810250 |
25 g | Plastic Bottle |
£29.40
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10497340
|
Thermo Scientific Chemicals
140811000 |
100 g | Plastic Bottle |
£71.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10716171
|
Thermo Scientific Chemicals
140815000 |
500 g | Plastic Bottle |
£206.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10697763
|
Thermo Scientific Chemicals
140810025 |
2.5 kg | Plastic Bottle |
£742.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
72-18-4 | |
117.15 | |
KZSNJWFQEVHDMF-BYPYZUCNSA-N | |
6287 | |
(2S)-2-amino-3-methylbutanoic acid |
C5H11NO2 | |
MFCD00064220 | |
l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin | |
CHEBI:16414 | |
CC(C)[C@H](N)C(O)=O |
Specifications
315.0°C | |
72-18-4 | |
200ppm max. | |
10ppm max. | |
C5H11NO2 | |
(CH3)2CHCH(NH2)CO2H | |
MFCD00064220 | |
13,226 | |
15, 10095 | |
Solubility in water: 85g/L (20°C). Other solubilities: practically insoluble in cold alcohol, ether and, acetone, soluble in mineral acids, insoluble in chloroform, diethylether | |
CC(C)[C@H](N)C(O)=O | |
+ 28.00 | |
6287 | |
300ppm max. | |
99% | |
1.2300g/mL | |
1.23 | |
L-Valine, 99% |
200ppm max. | |
White | |
0.5% max. | |
99% | |
Authentic | |
10ppm max. | |
04, 427 | |
0.2% max. | |
l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin | |
KZSNJWFQEVHDMF-BYPYZUCNSA-N | |
(2S)-2-amino-3-methylbutanoic acid | |
117.15 | |
CHEBI:16414 | |
117.15 | |
0.1% max. | |
+28.00 (20.00°C c=8,6N HCl) | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 200-773-6
missing translation for 'rtecsNumber' : YV9361000
missing translation for 'tsca' : TSCA
RUO – Research Use Only