Learn More
Thermo Scientific Chemicals L-Valine, 99%
£24.90 - £249.00
Chemical Identifiers
CAS | 72-18-4 |
---|---|
Molecular Formula | C5H11NO2 |
Molecular Weight (g/mol) | 117.15 |
MDL Number | MFCD00064220 |
InChI Key | KZSNJWFQEVHDMF-BYPYZUCNSA-N |
Synonym | l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin |
PubChem CID | 6287 |
ChEBI | CHEBI:16414 |
IUPAC Name | (2S)-2-amino-3-methylbutanoic acid |
SMILES | CC(C)[C@H](N)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11460653
|
Thermo Scientific Chemicals
A12720.14 |
25 g |
£24.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470653
|
Thermo Scientific Chemicals
A12720.22 |
100 g |
£63.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480653
|
Thermo Scientific Chemicals
A12720.36 |
500 g |
£249.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
L-Valine is used in the enhancement of muscle growth and energy. It is utilized in tissue repair. It is also involved in the studies to attenuate arrhythmias and induce hypotensive effects. It acts as a precursor for the preparation of penicillin through biosynthetic pathway. It also employed as nutritional supplements in food and beverage industries. Further, it plays an important role in maintaining mental vigor, muscle coordination and emotional calm.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water. Slightly soluble in methanol and acetone.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
72-18-4 | |
117.15 | |
KZSNJWFQEVHDMF-BYPYZUCNSA-N | |
6287 | |
(2S)-2-amino-3-methylbutanoic acid |
C5H11NO2 | |
MFCD00064220 | |
l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin | |
CHEBI:16414 | |
CC(C)[C@H](N)C(O)=O |
Specifications
∽315°C | |
99% | |
MFCD00064220 | |
14,9909 | |
Soluble in water. Slightly soluble in methanol and acetone. | |
CC(C)[C@H](N)C(O)=O | |
117.15 | |
CHEBI:16414 | |
99% | |
L-Valine |
72-18-4 | |
C5H11NO2 | |
1721136 | |
l-valine, valine, s-valine, s-2-amino-3-methylbutanoic acid, h-val-oh, 2-amino-3-methylbutyric acid, 2s-2-amino-3-methylbutanoic acid, s-2-amino-3-methylbutyric acid, l-alpha-amino-beta-methylbutyric acid, valinum latin | |
KZSNJWFQEVHDMF-BYPYZUCNSA-N | |
(2S)-2-amino-3-methylbutanoic acid | |
6287 | |
117.15 | |
1.23 |
Safety and Handling
missing translation for 'einecsNumber' : 200-773-6
missing translation for 'rtecsNumber' : YV9361000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only