missing translation for 'onlineSavingsMsg'
Learn More

L-Valinamide hydrochloride, 95%

Product Code. 15450649
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15450649 25 g 25g
15440649 5 g 5g
15460649 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15450649 Supplier Thermo Scientific Alfa Aesar Supplier No. H63775.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3014-80-0 | C5H13ClN2O | 152.62 g/mol

It is a reagent used in the synthesis of alkylpyrazines. Also used in the synthesis of elastase inhibitory activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reagent used in the synthesis of alkylpyrazines. Also used in the synthesis of elastase inhibitory activity

Solubility
Soluble in methanol (50 mg/ ml-clear, colorless solution).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3014-80-0
Molecular Formula C5H13ClN2O
Molecular Weight (g/mol) 152.62
MDL Number MFCD00039085
InChI Key XFCNYSGKNAWXFL-WCCKRBBISA-N
Synonym l-valinamide hydrochloride, h-val-nh2 hcl, h-val-nh2.hcl, l-valine amide hydrochloride, 2s-2-amino-3-methylbutanamide hydrochloride, 2s-2-amino-3-methylbutanamide, chloride, butanamide, 2-amino-3-methyl-, monohydrochloride, 2s, l-valine amide, h-d-val-nh2 hcl, d-valinamide-hcl
PubChem CID 13689583
IUPAC Name (2S)-2-amino-3-methylbutanamide;hydrochloride
SMILES Cl.CC(C)[C@H](N)C(N)=O

Specifications

Melting Point 266°C to 270°C
Beilstein 4198583
Solubility Information Soluble in methanol (50mg/ ml-clear,colorless solution).
Quantity 25 g
Formula Weight 152.62
Percent Purity 95%
Physical Form Solid
Chemical Name or Material L-Valinamide hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.