missing translation for 'onlineSavingsMsg'
Learn More

L-Valacyclovir Hydrochloride, TRC

Product Code. 30456282
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456282 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30456282 Supplier Toronto Research Chemicals Supplier No. TRCV085000100MG
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Valaciclovir hydrochloride
CAS 124832-27-5
Molecular Formula C13 H20 N6 O4 . Cl H
Quantity 100 mg
Synonym Valaciclovir hydrochloride anhydrous, 2-[(2-Amino-6-oxo-1,6-dihydro-9H-purin-9-yl)methoxy]ethyl l-valinate hydrochloride, L-Valine','2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]ethyl ester','monohydrochloride, 256U, 256U87 hydrochloride, BW 256, BW 256U87, Valaciclovir hydrochloride, Valacyclovir hydrochloride, Valtrex, Zelitrex, L-Valine','2-[(2-amino-1,6-dihydro-6-oxo-9H-purin-9-yl)methoxy]ethyl ester','hydrochloride (1:1), Aciclovir, GW277085A, 111U90, 4793W93, 23W91
InChI Formula InChI=1S/C13H20N6O4.ClH/c1-7(2)8(14)12(21)23-4-3-22-6-19-5-16-9-10(19)17-13(15)18-11(9)20;/h5,7-8H,3-4,6,14H2,1-2H3,(H3,15,17,18,20);1H/t8-;/m0./s1
SMILES Cl.CC(C)[C@H](N)C(=O)OCCOCn1cnc2C(=O)NC(=Nc12)N
IUPAC Name 2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]ethyl (2S)-2-amino-3-methylbutanoate;hydrochloride
Molecular Weight (g/mol) 360.8
Formula Weight 360.1313
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.