missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine, TRC

Product Code. 30008307
Change view
Click to view available options
Quantity:
100 mg
10 g
25 g
Unit Size:
100mg
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008307 10 g 10g
30006532 25 g 25g
17979534 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008307 Supplier Toronto Research Chemicals Supplier No. TRCT89997510G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-18-4
Molecular Formula C9 H11 N O3
Molecular Weight (g/mol) 181.19
Synonym L-Tyrosine, Tyrosine, L- (8CI), (-)-α-Amino-p-hydroxyhydrocinnamic acid, (2S)-2-Amino-3-(4-hydroxyphenyl)propanoic acid, (S)-2-Amino-3-(4-hydroxyphenyl)propanoic acid, (S)-Tyrosine, (S)-α-Amino-4-hydroxybenzenepropanoic acid, 4-(2-Amino-2-carboxyethyl)phenol, Benzenepropanoic acid, α-amino-4-hydroxy-, (S)-, H-L-Tyr-OH, L(-)-β-(p-Hydroxyphenyl)alanine, L-(-)-Tyrosine, L-4-Hydroxyphenylalanine, L-Phenylalanine, 4-hydroxy-, L-Tyr, L-p-Tyrosine, NSC 82624, NSC 9973, Propanoic acid, 2-amino-3-(4-hydroxyphenyl)-, (S)-, Therigon, Tyr, Tyrosine, p-Hydroxyphenylalanine, p-Tyrosine, β-(p-Hydroxyphenyl)alanine, Levodopa Imp. B (EP)
IUPAC Name (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid
SMILES N[C@@H](Cc1ccc(O)cc1)C(=O)O

Specifications

InChI Formula InChI=1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m0/s1
Quantity 10 g
Formula Weight 181.0739
Shipping Condition Room Temperature
Chemical Name or Material L-Tyrosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.