missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine, MP Biomedicals™

Product Code. 11487391 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
1 kg
Unit Size:
1kg
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487391 1 kg 1kg
11457391 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11487391 Supplier MP Biomedicals Supplier No. 0210318391
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 60-18-4 | C9H11NO3 | 181.19 g/mol

Chemical Identifiers

CAS 60-18-4
Molecular Formula C9H11NO3
Molecular Weight (g/mol) 181.19
MDL Number MFCD00002606
InChI Key OUYCCCASQSFEME-SVGMAFHSNA-N
Synonym l-tyrosine, tyrosine, s-tyrosine, p-tyrosine, h-tyr-oh, l-p-tyrosine, 2s-2-amino-3-4-hydroxyphenyl propanoic acid, 4-hydroxy-l-phenylalanine, l---tyrosine, tyrosine, l
PubChem CID 6057
ChEBI CHEBI:17895
IUPAC Name (2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C=C1)C(O)=O

Specifications

Color White
Assay Percent Range 0.97
Density 1.4g/mL
Quantity 1 kg
Formula Weight 181.19
Percent Purity 99.5%
Grade Research
Physical Form Powder
Chemical Name or Material L-Tyrosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.