missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Tyrosine Methyl Ester, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
L-Tyrosine Methyl Ester
Specifications
Specifications
| Chemical Name or Material | L-Tyrosine Methyl Ester |
| CAS | 1080-06-4 |
| Analyte or Component Names | L-Tyrosine Methyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H13 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | L-Tyrosine methyl ester, (S)-2-Amino-3-(4-hydroxyphenyl)propionic acid methyl ester, Methyl (2S)-2-amino-3-(4-hydroxyphenyl)propionate, Methyl (2S)-3-(4-hydroxyphenyl)-2-aminopropionate, Methyl (S)-2-amino-3-(4-hydroxyphenyl)propanoate, Methyl L-tyrosinate, Methyl tyrosinate, S-Tyrosine methyl ester, Tyrosine methyl ester |
| InChI Formula | InChI=1S/C10H13NO3/c1-14-10(13)9(11)6-7-2-4-8(12)5-3-7/h2-5,9,12H,6,11H2,1H3/t9-/m0/s1 |
| SMILES | COC(=O)[C@@H](N)Cc1ccc(O)cc1 |
| IUPAC Name | methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction