missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine methyl ester hydrochloride, 98+%

Product Code. 11435327
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11435327

Brand: Thermo Scientific Alfa Aesar B22683.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3417-91-2 | C10H14ClNO3 | 231.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3417-91-2
Molecular Formula C10H14ClNO3
Molecular Weight (g/mol) 231.68
MDL Number MFCD00012607
InChI Key VXYFARNRGZWHTJ-MVRPGTNWNA-N
Synonym l-tyrosine methyl ester hydrochloride, h-tyr-ome.hcl, methyl l-tyrosinate hydrochloride, h-tyr-ome hcl, methyl tyrosinate hydrochloride, unii-w42m0mi271, tyrosine methyl ester hydrochloride, methyl l-tyrosinate hcl, s-methyl 2-amino-3-4-hydroxyphenyl propanoate hydrochloride, s-2-amino-3-4-hydroxy-phenyl-propionic acid methyl ester hydrochloride
PubChem CID 76956
IUPAC Name methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate;hydrochloride
SMILES Cl.COC(=O)[C@@H](N)CC1=CC=C(O)C=C1

Specifications

Melting Point 187°C (decomposition)
Beilstein 3917353
Quantity 100 g
Formula Weight 231.68
Optical Rotation −5.2° (c= 2.4 in Water)
Sensitivity Hygroscopic
Percent Purity ≥98%
Chemical Name or Material L-Tyrosine methyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.