missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Tyrosine methyl ester, 98%

Product Code. 10061620 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10061620 100 g Glass bottle 100g
10022973 5 g Glass bottle 5g
10754851 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10061620 Supplier Thermo Scientific Chemicals Supplier No. 140651000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1080-06-4 | C10H13NO3 | 195.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1080-06-4
Molecular Formula C10H13NO3
Molecular Weight (g/mol) 195.22
MDL Number MFCD00002392
InChI Key MWZPENIJLUWBSY-VIFPVBQESA-N
Synonym l-tyrosine methyl ester, h-tyr-ome, tyrosine methyl ester, methyl l-tyrosinate, tyr-ome, methyl tyrosinate, tyrosine, methyl ester, s-2-amino-3-4-hydroxy-phenyl-propionic acid methyl ester, methyl 2s-2-amino-3-4-hydroxyphenyl propanoate, s-methyl 2-amino-3-4-hydroxyphenyl propanoate
PubChem CID 70652
ChEBI CHEBI:17215
IUPAC Name methyl (2S)-2-amino-3-(4-hydroxyphenyl)propanoate
SMILES COC(=O)[C@@H](N)CC1=CC=C(O)C=C1

Specifications

Melting Point 133°C to 135°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula 4-(HO)C6H4CH2CH(NH2)CO2CH3
Quantity 100 g
Beilstein 14, 612
Specific Rotation Condition + 25.00 (20.00°C c=2.4,CH3OH)
Specific Rotation + 25
Solubility Information Solubility in water: .. Other solubilities: 5% in methanol
Formula Weight 195.22
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Tyrosine methyl ester, 98%
Show More Show Less
compliance-icons
Product Identifier
  • L-Tyrosine methyl ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.