missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine, MedChemExpress

Product Code. 16492376
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
500 mg
Unit Size:
200mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492376 500 mg 500mg
16461506 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492376 Supplier Medchem Express Supplier No. HYN0473
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-Tyrosine is a non-essential amino acid which can inhibit citrate synthase activity in the posterior cortex.

L-Tyrosine is a non-essential amino acid which can inhibit citrate synthase activity in the posterior cortex.

Specifications

Chemical Name or Material L-Tyrosine
CAS 60-18-4
Color White
Purity Grade Notes Research
Molecular Formula C9H11NO3
Solubility Information 1M HCl : 50 mg/mL (275.95 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (5.52 mM; Need ultrasonic)
SMILES N[C@@H](CC1=CC=C(C=C1)O)C(O)=O
Molecular Weight (g/mol) 181.19
Quantity 500 mg
Formula Weight 181.19
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.