missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford L-Tyrosine, Disodium Salt
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- CAS Number: 122666-87-9
- Molecular Formula: C9H9NO3Na2 2H2O
- Molecular Weight: 262.19
Specifications
Specifications
| Chemical Name or Material | L-Tyrosine, Disodium Salt |
| CAS | 122666-87-9 |
| Molecular Formula | C9H9NO3Na2-2H2O |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction