missing translation for 'onlineSavingsMsg'
Learn More

L-Tryptophan, TRC

Product Code. 17967904
Change view
Click to view available options
Quantity:
10 mg
10 g
25 g
Unit Size:
10g
10mg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17967904 25 g 25g
17947444 10 g 10g
17959074 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17967904 Supplier Toronto Research Chemicals Supplier No. TRCT94721025G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-22-3
Molecular Formula C11 H12 N2 O2
Molecular Weight (g/mol) 204.23
Synonym L-Tryptophan, (-)-Tryptophan, (2S)-2-Amino-3-(1H-indol-3-yl)propanoic acid, (S)-α-Amino-β-indolepropionic acid, (S)-α-Amino-1H-indole-3-propanoic acid, (S)-α-Aminoindole-3-propionic acid, (S)-Tryptophan, 1H-Indole-3-alanine, (S)-, 1H-Indole-3-propanoic acid, α-amino-, (S)-, 2-Amino-3-indolylpropanoic acid, 3-Indol-3-ylalanine, EH 121, L-(-)-Tryptophan, 3-(1H-Indol-3-yl)-L-alanine, L-Tryptophane, Lyphan, NSC 13119, Secretory peptide (human clone HCQCJ56 precursor), Tryptophan, Tryptophane, h-Trp-oh, l-α-Aminoindole-3-propionic acid, l-β-3-Indolylalanine
IUPAC Name (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES N[C@@H](Cc1c[nH]c2ccccc12)C(=O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13H,5,12H2,(H,14,15)/t9-/m0/s1
Formula Weight 204.0899
Percent Purity >95%
Chemical Name or Material L-(-)-Tryptophan
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.