missing translation for 'onlineSavingsMsg'
Learn More

L-Tryptophan, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15508894 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15508894 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15508894 Supplier Avantor J.T.Baker Supplier No. 209206

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 73-22-3
Molecular Formula C11H12N2O2
Packaging Poly Bottle
MDL Number MFCD00064340
Synonym l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
InChI Key QIVBCDIJIAJPQS-VIFPVBQESA-N
SMILES N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
IUPAC Name (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
Molecular Weight (g/mol) 204.23
Quantity 1 kg
PubChem CID 6305
ChEBI CHEBI:16828
Formula Weight 204.23
Specific Gravity 1L = 1.34kg
Grade Multi-Compendial/USP
Chemical Name or Material L-Tryptophan
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.