missing translation for 'onlineSavingsMsg'
Learn More

L-Tryptophan Methyl Ester Hydrochloride, Mikromol

Product Code. 30462079
Change view
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462079 4 x 25 mg -
30052161 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30462079 Supplier Mikromol Supplier No. MM0172.1500254AMP
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

L-Tryptophan Methyl Ester Hydrochloride, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Tryptophan Methyl Ester Hydrochloride
CAS 7524-52-9
Analyte or Component Names L-Tryptophan Methyl Ester Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C12 H14 N2 O2 . Cl H
Certifications/Compliance ISO 17025
Synonym L-Tryptophan methyl ester, hydrochloride (1:1), L-Tryptophan, methyl ester, monohydrochloride (9CI), Tryptophan, methyl ester, monohydrochloride, L- (6CI,7CI,8CI), (S)-Tryptophan methyl ester hydrochloride, (S)-Tryptophan methyl ester monohydrochloride, L-(-)-Tryptophan methyl ester hydrochloride, L-Tryptophan methyl ester hydrochloride, Methyl L-tryptophanate hydrochloride, Tryptophan methyl ester hydrochloride, Tryptophan methyl ester monohydrochloride
InChI Formula InChI=1S/C12H14N2O2.ClH/c1-16-12(15)10(13)6-8-7-14-11-5-3-2-4-9(8)11;/h2-5,7,10,14H,6,13H2,1H3;1H/t10-;/m0./s1
Solution Type Neat
SMILES Cl.COC(=O)[C@@H](N)Cc1c[nH]c2ccccc12
IUPAC Name methyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate;hydrochloride
Molecular Weight (g/mol) 254.71
Quantity 4 x 25 mg
Formula Weight 254.0822 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.