missing translation for 'onlineSavingsMsg'
Learn More

L-Tryptophan methyl ester hydrochloride, 98%

Product Code. 10230373 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10230373 25 g Glass Bottle 25g
10493063 5 g Plastic Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10230373 Supplier Thermo Scientific Acros Supplier No. 376650250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7524-52-9 | C12H15ClN2O2 | 254.71 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7524-52-9
Molecular Formula C12H15ClN2O2
Molecular Weight (g/mol) 254.71
MDL Number MFCD00066134
InChI Key XNFNGGQRDXFYMM-PPHPATTJSA-N
Synonym l-tryptophan methyl ester hydrochloride, h-trp-ome.hcl, methyl l-tryptophanate hydrochloride, methyl tryptophanate hydrochloride, methyl l-tryptophanate hcl, unii-au62o6861l, methyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride, s-methyl 2-amino-3-1h-indol-3-yl propanoate hydrochloride, l-tryptophanmethylesterhydrochloride, l-tryptophan methyl ester hcl
PubChem CID 2734891
IUPAC Name methyl (2S)-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride
SMILES Cl.COC(=O)[C@@H](N)CC1=CNC2=CC=CC=C12

Specifications

Melting Point 218.0°C to 220.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Specific Rotation + 18.00
Specific Rotation Condition + 18.00 (20.00°C c=5 CH3OH)
Quantity 25 g
Formula Weight 254.72
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Tryptophan methyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.