missing translation for 'onlineSavingsMsg'
Learn More

L-Tryptophan, MedChemExpress

Product Code. 16449116
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16449116 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16449116 Supplier Medchem Express Supplier No. HYN0623
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-Tryptophan (Tryptophan) is an essential amino acid that is the precursor of serotonin, melatonin, and vitamin B3.

L-Tryptophan (Tryptophan) is an essential amino acid that is the precursor of serotonin, melatonin, and vitamin B3.

Specifications

Chemical Name or Material L-Tryptophan
CAS 73-22-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H12N2O2
Synonym Tryptophan Tryptophane
Solubility Information H2O : 12.5 mg/mL (61.21 mM; Need ultrasonic) ∣DMSO : 7.69 mg/mL (37.65 mM; Need ultrasonic)
SMILES N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
Molecular Weight (g/mol) 204.23
Quantity 100 mg
Formula Weight 204.23
Percent Purity 99.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.