missing translation for 'onlineSavingsMsg'
Learn More

L-Tryptophan, Extra Pure, Fisher Chemical

Product Code. 10448230 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10448230 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10448230 Supplier Fisher Chemical Supplier No. T/3770/46
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

25g, Plastic powder jar

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 280°C
CAS 73-22-3
pH 5.5 to 7.0
Color White
Molecular Formula C11H12N2O2
Packaging Plastic Powder Jar
MDL Number MFCD00064340
Synonym l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
InChI Key QIVBCDIJIAJPQS-VIFPVBQESA-N
SMILES N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O
IUPAC Name (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
Molecular Weight (g/mol) 204.23
Quantity 25 g
PubChem CID 6305
ChEBI CHEBI:16828
Formula Weight 204.23g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.