missing translation for 'onlineSavingsMsg'
Learn More

L-Tryptophan benzyl ester hydrochloride, 98%, Thermo Scientific™

Product Code. 15490049
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
L-Tryptophan benzyl ester hydrochloride is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol (50 mg/mL in MeOH).

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35858-81-2
Molecular Formula C18H18ClN2O2
Molecular Weight (g/mol) 329.80
MDL Number MFCD00058122
InChI Key BYXCCQMIJIPTSL-UHFFFAOYNA-N
Synonym h-trp-obzl.hcl, l-tryptophan benzyl ester hydrochloride, benzyl l-tryptophanate hydrochloride, h-trp-obzl hcl, h-trp-obzl hydrochloride, s-benzyl 2-amino-3-1h-indol-3-yl propanoate hydrochloride, h-trp-obzlhcl, benzyl 2s-2-amino-3-1h-indol-3-yl propanoate hydrochloride, benzyl l-tryptophanate monohydrochloride, c18h18n2o2.hcl
PubChem CID 45073232
SMILES [Cl].NC(CC1=CNC2=CC=CC=C12)C(=O)OCC1=CC=CC=C1

Specifications

Color White
Solubility Information Soluble in methanol (50mg/mL in MeOH).
IUPAC Name benzyl 2-amino-3-(1H-indol-3-yl)propanoate hydrochloridyl
Formula Weight 330.81
Percent Purity 98%
Quantity 1 g
Physical Form Powder
Chemical Name or Material L-Tryptophan benzyl ester hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'