missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(-)-Tryptophan, 99%

Product Code. 10316980 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
100g
1kg
25g
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10316980 25 g Plastic Bottle 25g
10306980 5 g Glass Bottle 5g
10543881 100 g Plastic Bottle 100g
10193520 500 g Plastic Bottle 500g
10544264 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10316980 Supplier Thermo Scientific Chemicals Supplier No. 140590250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L(-)-Tryptophan, CAS # 73-22-3, is an essential amino acid necessary for the biosynthesis of proteins. | CAS: 73-22-3 | C11H12N2O2 | 204.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • L(-)-Tryptophan is a nonpolar aromatic compound and essential amino acid required for protein synthesis
  • It is a biochemical precursor of niacin, indole alkaloids, and serotonin

Applications

  • L-Tryptophan acts as a natural dietary supplement and has laboratory applications in the research of antidepressants, anxiolytics, and sleep aids
  • It is a precursor of metabolites that are important in the laboratory study of mammalian physiology, including gastrointestinal functions, immunity, metabolism, and the nervous system
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-22-3
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
MDL Number MFCD00064340
InChI Key QIVBCDIJIAJPQS-VIFPVBQESA-N
Synonym l-tryptophan, tryptophan, l-tryptophane, s-tryptophan, tryptophane, h-trp-oh, optimax, trofan, tryptacin, ardeytropin
PubChem CID 6305
ChEBI CHEBI:16828
IUPAC Name (2S)-2-amino-3-(1H-indol-3-yl)propanoic acid
SMILES N[C@@H](CC1=CNC2=CC=CC=C12)C(O)=O

Specifications

Melting Point 280.0°C to 285.0°C
Color White to White-Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Loss on Drying 0.5% max. (1 g, 105°C)
Packaging Plastic Bottle
Beilstein 22, 546
Specific Rotation − 31.10
Merck Index 15, 9977
Solubility Information Solubility in water: 11.4g/L (25°C). Other solubilities: soluble in alkali hydroxides and hot alcohol, insoluble in chloroform
Specific Rotation Condition − 31.10 (20.00°C c=1,H20)
Quantity 25 g
Formula Weight 204.23
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material L(-)-Tryptophan, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.