missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ L-Thyroxine T4 Monoclonal Antibody (T4YCH)

Description
MIT0501PG targets L-Thyroxine (T4) in ELISA, IP, and RIA applications and shows reactivity with Chemical samples. The MIT0501PG immunogen is thyroxine-BSA. MIT0501PG was formerly sold as a Seradyn product.
Levothyroxine, also L-thyroxine, synthetic T4, or 3,5,3',5'-tetraiodo-L-thyronine, is a synthetic form of thyroxine (thyroid hormone), used as a hormone replacement for patients with thyroid problems. The natural hormone is chemically in the chiral L-form, as is the pharmaceutical agent. Dextrothyroxine (D-thyroxine) briefly saw research as an anticholesterol agent but was pulled due to cardiac side-effects.
Specifications
Specifications
| Antigen | L-Thyroxine T4 |
| Applications | ELISA, Immunoprecipitation, Radioimmune Assays (RIA) |
| Classification | Monoclonal |
| Clone | T4YCH |
| Concentration | 2.76 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.09% sodium azide; pH 7.4 |
| Gene Alias | C15H11I4NO4; Levothyroxine; L-thyroxine; Synthroid; Thyroxine |
| Host Species | Mouse |
| Immunogen | Thyroxine-BSA. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?