missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Thyroxine sodium salt pentahydrate, MedChemExpress
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
Description
L-Thyroxine sodium salt pentahydrate (Levothyroxine
Specifications
Specifications
| Chemical Name or Material | L-Thyroxine sodium salt pentahydrate |
| CAS | 6106-07-6 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C15H20I4NNaO9 |
| Quantity | 500 mg |
| Synonym | Sodium levothyroxine pentahydrate |
| Solubility Information | DMSO : ≥ 28 mg/mL (31.50 mM) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | N[C@@H](CC1=CC(I)=C(C(I)=C1)OC2=CC(I)=C(O)C(I)=C2)C(O[Na])=O.O.O.O.O.O |
| Molecular Weight (g/mol) | 888.93 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction