missing translation for 'onlineSavingsMsg'
Learn More

L-Thyroxine, MedChemExpress

Product Code. 16441896
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16441896 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16441896 Supplier Medchem Express Supplier No. HY18341
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress L-Thyroxine (Levothyroxine; T4) is a synthetic hormone for the research of hypothyroidism. DIO enzymes convert biologically active thyroid hormone (Triiodothyronine,T3) from L-Thyroxine (T4).

L-Thyroxine (Levothyroxine

Specifications

Chemical Name or Material L-Thyroxine
CAS 51-48-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C15H11I4NO4
Quantity 500 mg
Synonym Levothyroxine T4
Solubility Information DMSO : 250 mg/mL (321.80 mM; Need ultrasonic)
SMILES N[C@@H](CC1=CC(I)=C(C(I)=C1)OC2=CC(I)=C(O)C(I)=C2)C(O)=O
Molecular Weight (g/mol) 776.87
Formula Weight 776.87
Percent Purity 96.76%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.