missing translation for 'onlineSavingsMsg'
Learn More

L-Threoninol, TRC

Product Code. 30454191
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30454191 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454191 Supplier Toronto Research Chemicals Supplier No. TRCT4055781G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Threoninol
CAS 3228-51-1
Molecular Formula C4H11NO2
Quantity 1 g
InChI Formula InChI=1S/C4H11NO2/c1-3(7)4(5)2-6/h3-4,6-7H,2,5H2,1H3/t3-,4-/m1/s1
SMILES C[C@@H](O)[C@H](N)CO
IUPAC Name (2R,3R)-2-aminobutane-1,3-diol
Molecular Weight (g/mol) 105.14
Formula Weight 105.079
Product Title
Select an issue

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.