missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Threonine, 98+%

Product Code. 11452878
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452878 500 g 500g
11432878 25 g 25g
11442878 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452878 Supplier Thermo Scientific Chemicals Supplier No. A16851.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 72-19-5 | C4H9NO3 | 119.12 g/mol

L-Threonine is used to alleviate anxiety and mild depression. It is an important component of many proteins such as tooth enamel, collagen, and elastin. It is an important amino acid in the nervous system and plays a vital role in porphyrin and fat metabolism. It is useful in intestinal disorders and indigestion. It is also used as a prodrug to reliably elevate brain glycine levels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Threonine is used to alleviate anxiety and mild depression. It is an important component of many proteins such as tooth enamel, collagen, and elastin. It is an important amino acid in the nervous system and plays a vital role in porphyrin and fat metabolism. It is useful in intestinal disorders and indigestion. It is also used as a prodrug to reliably elevate brain glycine levels.

Solubility
Soluble in water. Insoluble in ethanol, ethyl ether and chloroform.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72-19-5
Molecular Formula C4H9NO3
Molecular Weight (g/mol) 119.12
MDL Number MFCD00064270
InChI Key AYFVYJQAPQTCCC-UHFFFAOYNA-N
Synonym l-threonine, threonine, threonin, h-thr-oh, 2s,3r-2-amino-3-hydroxybutanoic acid, s-threonine, l---threonine, 2-amino-3-hydroxybutyric acid, threonine van, threoninum latin
PubChem CID 6288
ChEBI CHEBI:16857
SMILES CC(O)C(N)C(O)=O

Specifications

Melting Point 255°C (decomposition)
Beilstein 1721646
Merck Index 14,9380
Solubility Information Soluble in water. Insoluble in ethanol,ethyl ether and chloroform.
IUPAC Name 2-amino-3-hydroxybutanoic acid
Quantity 500 g
Formula Weight 119.12
Optical Rotation −28° (c=5 in Water)
Percent Purity ≥98%
Chemical Name or Material L-Threonine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.