missing translation for 'onlineSavingsMsg'
Learn More

L-threo-Hex-2-enaric acid 1,4-Lactone 6-Methyl Ester, TRC

Product Code. 17909744
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17909744 2.5 mg 2.5mg
30008802 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17909744 Supplier Toronto Research Chemicals Supplier No. TRCH2948352.5MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122046-79-1
Molecular Formula C7 H8 O7
Molecular Weight (g/mol) 204.13
Synonym L-threo-Hex-2-enaric acid, 1,4-lactone, 6-methyl ester (9CI), Methyl (R)-[(2R)-3,4-Dihydroxy-5-oxo-2,5-dihydrofuran-2-yl]hydroxyacetate
IUPAC Name methyl (2R)-2-[(2R)-3,4-dihydroxy-5-oxo-2H-furan-2-yl]-2-hydroxyacetate
SMILES COC(=O)[C@H](O)[C@H]1OC(=O)C(=C1O)O

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C7H8O7/c1-13-6(11)4(10)5-2(8)3(9)7(12)14-5/h4-5,8-10H,1H3/t4-,5+/m1/s1
Formula Weight 204.027
Percent Purity >95%
Chemical Name or Material L-threo-Hex-2-enaric acid 1,4-Lactone 6-Methyl ester
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.