missing translation for 'onlineSavingsMsg'
Learn More

L-Thiazolidine-4-carboxylic acid, 98%

Product Code. 11497277
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11497277 10 g 10g
11407287 50 g 50g
11417287 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497277 Supplier Thermo Scientific Alfa Aesar Supplier No. A15033.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 34592-47-7 | C4H7NO2S | 133.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34592-47-7
Molecular Formula C4H7NO2S
Molecular Weight (g/mol) 133.165
MDL Number MFCD00005212
InChI Key DZLNHFMRPBPULJ-VKHMYHEASA-N
Synonym l-thioproline, r-thiazolidine-4-carboxylic acid, l-thiaproline, l-thiazolidine-4-carboxylic acid, 4r-1,3-thiazolidine-4-carboxylic acid, h-thz-oh, r-4-thiazolidinecarboxylic acid, 4-thiaproline, thioproline, gamma-thioproline
PubChem CID 93176
ChEBI CHEBI:45171
IUPAC Name (4R)-1,3-thiazolidine-4-carboxylic acid
SMILES C1C(NCS1)C(=O)O

Specifications

Melting Point 192°C to 200°C (decomposition)
Quantity 10 g
UN Number UN2811
Beilstein 81065
Merck Index 14,9445
Optical Rotation −141° (c=1.3 in Water)
Formula Weight 133.17
Percent Purity 98%
Chemical Name or Material L-Thiazolidine-4-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.