missing translation for 'onlineSavingsMsg'
Learn More

L-Theanine, 98%

Product Code. 15580624 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15580624

Brand: Thermo Scientific Acros 459340050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3081-61-6 | C7H14N2O3 | 174.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3081-61-6
Molecular Formula C7H14N2O3
Molecular Weight (g/mol) 174.20
MDL Number MFCD00059653
InChI Key DATAGRPVKZEWHA-UHFFFAOYNA-N
Synonym l-theanine, theanine, theanin, s-2-amino-5-ethylamino-5-oxopentanoic acid, n-ethyl-l-glutamine, suntheanine, n5-ethyl-l-glutamine, n 5-ethyl-l-glutamine, unii-8021pr16qo
PubChem CID 439378
ChEBI CHEBI:17394
IUPAC Name (2S)-2-amino-5-(ethylamino)-5-oxopentanoic acid
SMILES CCNC(=O)CCC(N)C(O)=O

Specifications

Melting Point 206.0°C to 219.0°C
Infrared Spectrum Authentic
Linear Formula CH3CH2NHC(=O)CH2CH2CH(NH2)C(=O)OH
Quantity 5 g
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in ether, alcohols
Formula Weight 174.2
Percent Purity 98%
Chemical Name or Material L-Theanine
compliance-icons
Product Identifier
  • L-Theanine
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1B
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.