missing translation for 'onlineSavingsMsg'
Learn More

L-tert-Leucine, 99%

Product Code. 10260903 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10260903 1 g Glass Bottle 1g
10339260 5 g Glass Bottle 5g
10267353 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10260903 Supplier Thermo Scientific Acros Supplier No. 297780010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 20859-02-3 | C6H13NO2 | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20859-02-3
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064218
InChI Key NPDBDJFLKKQMCM-SGAVLPGINA-N
Synonym l-tert-leucine, 2s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valine, l-valine,3-methyl, h-tle-oh, s-2-amino-3,3-dimethylbutanoic acid, 3-methyl-l-valin, l-2-amino-3,3-dimethylbutanoic acid, s-2-amino-3,3-dimethylbutyric acid
PubChem CID 164608
IUPAC Name (2S)-2-amino-3,3-dimethylbutanoic acid
SMILES CC(C)(C)[C@H](N)C(O)=O

Specifications

Melting Point >300.0°C
Color White
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 99%
Loss on Drying 0.2% max. (105°C, 3 hrs)
Packaging Glass Bottle
Specific Rotation + 6.30
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 125.5g/L (20°C).
Specific Rotation Condition +6.3° (200°C c=4,6N HCl)
Quantity 1 g
Formula Weight 131.17
Percent Purity 99%
Physical Form Powder
Chemical Name or Material L-tert-Leucine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.