missing translation for 'onlineSavingsMsg'
Learn More

L+-Tartaric Acid, Disodium Salt, Dihydrate, Reagent ACS (crystals), ≥99%, Thermo Scientific Chemicals

Product Code. 13468578
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6106-24-7
Molecular Formula C4H8Na2O8
Molecular Weight (g/mol) 230.08
MDL Number MFCD00150035
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
Synonym unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi
PubChem CID 131855972
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

pH 7.0 to 9.0 (5% soln. at 25°C)
Color White
Assay Percent Range 99+%
Linear Formula (CHOHCOONa)2·2H2O
Solubility Information Solubility in water: 290g/L (20°C). Other solubilities: insolubel in organic solvents
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate
Formula Weight 230.08
Percent Purity ≥99%
Grade ACS Reagent
Quantity 10 kg
Infrared Spectrum Authentic
Loss on Drying 15.61% to 15.71% (150°C)
Packaging Plastic drum
Physical Form Needles or Powder
Chemical Name or Material L(+)-Tartaric acid, disodium salt, dihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'