missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L(+)-Glutamine, 99%
£38.20 - £487.00
Chemical Identifiers
CAS | 56-85-9 |
---|---|
Molecular Formula | C5H10N2O3 |
Molecular Weight (g/mol) | 146.15 |
MDL Number | MFCD00008044 |
InChI Key | ZDXPYRJPNDTMRX-VKHMYHEASA-N |
Synonym | l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid |
PubChem CID | 5961 |
ChEBI | CHEBI:18050 |
IUPAC Name | (2S)-2,5-diamino-5-oxopentanoic acid |
SMILES | N[C@@H](CCC(N)=O)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10376840
|
Thermo Scientific Chemicals
119951000 |
100 g | Plastic Bottle |
£38.20
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10645732
|
Thermo Scientific Chemicals
119955000 |
500 g | Plastic Bottle |
£135.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10328843
|
Thermo Scientific Chemicals
119950025 |
2.5 kg | Plastic Bottle |
£487.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
56-85-9 | |
146.15 | |
ZDXPYRJPNDTMRX-VKHMYHEASA-N | |
5961 | |
(2S)-2,5-diamino-5-oxopentanoic acid |
C5H10N2O3 | |
MFCD00008044 | |
l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid | |
CHEBI:18050 | |
N[C@@H](CCC(N)=O)C(O)=O |
Specifications
185.0°C | |
4 to 6 (2.5% soln. at 25°C) | |
200ppm max. | |
99% | |
Authentic | |
10ppm max. | |
04, 491 | |
15, 4507 | |
l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid | |
ZDXPYRJPNDTMRX-VKHMYHEASA-N | |
(2S)-2,5-diamino-5-oxopentanoic acid | |
146.15 | |
CHEBI:18050 | |
146.15 | |
99% | |
Crystalline Powder or Crystals |
56-85-9 | |
White | |
10ppm max. | |
C5H10N2O3 | |
H2NCOCH2CH2CH(NH2)CO2H | |
MFCD00008044 | |
0.2% max. (105°C, 3 hrs) | |
0.5% max. (TLC) | |
Solubility in water: 35g/L (20°C). Other solubilities: practically insoluble in methanol, ethanol, ether,, benzene, acetone, ethyl acetate, chloroform, soluble in mineral acids, soluble in ethanol: 46 μg/100mL (25°C), soluble in methanol: 3.5 mg/100mL (2 | |
N[C@@H](CCC(N)=O)C(O)=O | |
+31.5° to +33° (20°C, 589nm) (c=10, 2N HCl) | |
5961 | |
200ppm max. | |
95% min. (c=10, 2N HCl, 430nm) 1cm cell | |
0.1% max. | |
L(+)-Glutamine, 99% |
Safety and Handling
MOISTURE SENSITIVE
LIGHT SENSITIVE
missing translation for 'einecsNumber' : 200-292-1
missing translation for 'rtecsNumber' : MA2275100
missing translation for 'tsca' : TSCA
RUO – Research Use Only