missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Sorbose, ≥98%, For HPLC analysis, MP Biomedicals™

Product Code. p-4404535 Shop All MP Biomedicals Products
Click to view available options
Quantity:
500 g
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 11436352

Brand: MP Biomedicals 0210293990

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 87-79-6
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
InChI Key BJHIKXHVCXFQLS-OTWZMJIISA-N
Synonym l---sorbose, 3s,4r,5s-1,3,4,5,6-pentahydroxyhexan-2-one, sorbinose, keto-l-sorbose, esorben, sorbin, l--sorbose, hsdb 780, l-sorbinose, l-xylo-2-hexulose
PubChem CID 6904
ChEBI CHEBI:13172
IUPAC Name (3S,4R,5S)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Color White to Yellow
Quantity 500 g
Formula Weight 180.2
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material L-(-)-Sorbose, For HPLC analysis
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.