missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Sorbose, 98%

Product Code. 10165303 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10165303 1 kg Plastic bottle 1kg
10441841 100 g Plastic bottle 100g
10270943 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10165303 Supplier Thermo Scientific Acros Supplier No. 225960010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 87-79-6 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-79-6
MDL Number MFCD00151097
InChI Key BJHIKXHVCXFQLS-OTWZMJIISA-N
Synonym l---sorbose, 3s,4r,5s-1,3,4,5,6-pentahydroxyhexan-2-one, sorbinose, keto-l-sorbose, esorben, sorbin, l--sorbose, hsdb 780, l-sorbinose, l-xylo-2-hexulose
PubChem CID 6904
ChEBI CHEBI:13172
IUPAC Name (3S,4R,5S)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O

Specifications

Melting Point 158°C to 160°C
Color White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Plastic bottle
Molecular Formula C6H12O6
Quantity 1 kg
Merck Index 15,8852
Specific Rotation Condition - 43.00 (20.00°C c=5,H2O,23 C)
Specific Rotation -43
Solubility Information Solubility in water: soluble. Other solubilities: almost insoluble in alcohol,slightly soluble in ethanol,ethylether,methanol
Molecular Weight (g/mol) 180.16
Formula Weight 180.16
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material L(-)-Sorbose, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.