missing translation for 'onlineSavingsMsg'
Learn More

L-Serine, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 12614735 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12614735 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12614735 Supplier Avantor J.T.Baker Supplier No. 208806

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 56-45-1
Molecular Formula C3H7NO3
Packaging Poly Bottle
Synonym l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
InChI Key MTCFGRXMJLQNBG-REOHCLBHSA-N
SMILES C(C(C(=O)O)N)O
IUPAC Name (2S)-2-amino-3-hydroxypropanoic acid
Molecular Weight (g/mol) 105.093
Quantity 1 kg
PubChem CID 5951
ChEBI CHEBI:17115
Formula Weight 105.09
Grade Multi-Compendial/USP
Chemical Name or Material L-Serine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.