missing translation for 'onlineSavingsMsg'
Learn More

L-Serine, MP Biomedicals™

Product Code. 11446252 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446252 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11446252 Supplier MP Biomedicals Supplier No. 0210287391
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Functions as a precursor of glycine by serine hydroxymethyl transferase. L-Serine, MP Biomedicals is one of the two biological amino acids with a hydroxyl substituted side chain. It is hydrophilic in character.

L-serine is used in the study of influence of pulsed applications of serine on polyglucose synthesis by Fusobacterium nucleatum cultures. Supplementation of minimal media with serine has been shown to restore growth in Escherichia coli that lack the PII and GlnK genes.

Specifications

CAS 56-45-1
Melting Point 222°C (decomposition)(literature)
Molecular Formula C3H7NO3
Quantity 1 kg
Synonym l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
Solubility Information Soluble in water (50mg/mL); insoluble in alcohol and ether.
InChI Key MTCFGRXMJLQNBG-REOHCLBHSA-N
Optical Rotation 10–20° (c=10, 2N HCl)
SMILES C(C(C(=O)O)N)O
IUPAC Name (2S)-2-amino-3-hydroxypropanoic acid
Molecular Weight (g/mol) 105.093
PubChem CID 5951
ChEBI CHEBI:17115
Formula Weight 105.09
Percent Purity ≥99%
Physical Form Free-flowing Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.