missing translation for 'onlineSavingsMsg'
Learn More

L-Serine ethyl ester hydrochloride, 99%

Product Code. 11482227
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11482227 25 g 25g
11472227 5 g 5g
11492227 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11482227 Supplier Thermo Scientific Alfa Aesar Supplier No. A14174.14
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 26348-61-8 | C5H12ClNO3 | 169.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Serine ethyl ester hydrochloride is widely used in pharmaceutical and food industry.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents and moisture. Store under dry inert gas. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26348-61-8
Molecular Formula C5H12ClNO3
Molecular Weight (g/mol) 169.61
MDL Number MFCD00012594
InChI Key JZJQCLZQSHLSFB-UHFFFAOYNA-N
Synonym l-serine ethyl ester hydrochloride, h-ser-oet.hcl, ethyl l-serinate hydrochloride, s-ethyl 2-amino-3-hydroxypropanoate hydrochloride, serine ethyl ester hydrochloride, l-serine ethyl ester hcl, l-serine, ethyl ester, hydrochloride, ethyl serinate hydrochloride, ethyl 2s-2-amino-3-hydroxypropanoate hydrochloride, h-ser-oet hcl
PubChem CID 2724185
SMILES [H+].[Cl-].CCOC(=O)C(N)CO

Specifications

Melting Point 130°C to 132°C
Beilstein 3562346
Solubility Information Soluble in water.
IUPAC Name ethyl (2S)-2-amino-3-hydroxypropanoate;hydrochloride
Quantity 25 g
Formula Weight 169.61
Optical Rotation −4.4° (c=4 in Water)
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material L-Serine ethyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.