missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Serine, Cell Culture Reagent

Product Code. 15414609
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15414609 100 g 100g
15404609 10 g 10g
15424609 500 g 500g
15434609 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15414609 Supplier Thermo Scientific Chemicals Supplier No. J62187.22
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56-45-1 | C3H7NO3 | 105.093 g/mol

It is a precursor of glycine by serine hydroxymethyltransferase. D-serine, synthesized by serine racemase from L-serine, serves as a neuronal signaling molecule by activating NMDA receptors in the brain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a precursor of glycine by serine hydroxymethyltransferase. D-serine, synthesized by serine racemase from L-serine, serves as a neuronal signaling molecule by activating NMDA receptors in the brain.

Solubility
Soluble in water.

Notes
Store in cool, dry place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-45-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00064224
InChI Key MTCFGRXMJLQNBG-REOHCLBHSA-N
Synonym l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
PubChem CID 5951
ChEBI CHEBI:17115
IUPAC Name (2S)-2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O

Specifications

Melting Point 222°C
Color White
Beilstein 1721404
Merck Index 14,8460
Solubility Information Soluble in water.
Quantity 100 g
Formula Weight 105.09
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Serine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.