missing translation for 'onlineSavingsMsg'
Learn More

L-Serine-2,3,3-d3, TRC

Product Code. 30135447
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135447 50 mg 50mg
17948369 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30135447 Supplier Toronto Research Chemicals Supplier No. TRCS27099250MG
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105591-10-4
Molecular Formula C3 D3 H4 N O3
Molecular Weight (g/mol) 108.1111
Synonym L-Serine-2,3,3-d3 (9CI, ACI), (2S)-2-Amino-3-hydroxypropanoic-2,3,3-d3 acid, L-Serine-2,3,3-D3
IUPAC Name (2S)-2-amino-2,3,3-trideuterio-3-hydroxypropanoic acid
SMILES [2H]C([2H])(O)[C@]([2H])(N)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)/t2-/m0/s1/i1D2,2D
Formula Weight 108.0614
Chemical Name or Material L-Serine-2,3,3-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.