missing translation for 'onlineSavingsMsg'
Learn More

L-SelenoMethionine, MedChemExpress

Product Code. 16462236
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462236 500 mg 500mg
16428556 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16462236 Supplier Medchem Express Supplier No. HYB1000A
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-SelenoMethionine, an L-isomer of Selenomethionine, is a major natural food-form of selenium. L-SelenoMethionin is a cancer chemopreventive agent that can reduce cancer incidence by dietary supplementation and induce apoptosis of cancer cells. L-SelenoMethionine also can increase expression of glutathione peroxidase.

L-SelenoMethionine, an L-isomer of Selenomethionine, is a major natural food-form of selenium. L-SelenoMethionin is a cancer chemopreventive agent that can reduce cancer incidence by dietary supplementation and induce apoptosis of cancer cells. L-SelenoMethionine also can increase expression of glutathione peroxidase.

Specifications

Chemical Name or Material L-SelenoMethionine
CAS 3211-76-5
Color White
Purity Grade Notes Research
Molecular Formula C5H11NO2Se
Solubility Information H2O : 14.29 mg/mL (72.87 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (5.10 mM; Need ultrasonic)
SMILES O=C(O)[C@@H](N)CC[Se]C
Molecular Weight (g/mol) 196.11
Quantity 500 mg
Formula Weight 196.11
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.