missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Ribose, 99%

Product Code. 15550524 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15550524 1 g 1g
15560524 5 g 5g
15570524 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15550524 Supplier Thermo Scientific Acros Supplier No. 257910010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24259-59-4 | C5H10O5 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24259-59-4
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00167010
InChI Key HMFHBZSHGGEWLO-ZHZWXSSZNA-N
Synonym l-ribose, l-+-ribose, 2s,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-ribose, aldehydo-l-ribo-pentose, ror, l-ribose 9ci, ksc202k1p
PubChem CID 90428
ChEBI CHEBI:47015
IUPAC Name (2S,3S,4S)-2,3,4,5-tetrahydroxypentanal
SMILES OC[C@@H]1OC(O)[C@@H](O)[C@H]1O

Specifications

Melting Point 84.0°C to 86.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Packaging Glass Bottle
Quantity 1 g
Beilstein 01,859
Specific Rotation Condition + 20.00 (20.00°C c=2,water)
Specific Rotation + 20.00
Solubility Information (10% in water) Clear colorless
Formula Weight 150.13
Percent Purity 99%
Chemical Name or Material L(+)-Ribose
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.