missing translation for 'onlineSavingsMsg'
Learn More

α-L-Rhamnose monohydrate, MedChemExpress

Product Code. 16469116
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16469116 100 mg 100mg
16452386 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16469116 Supplier Medchem Express Supplier No. HYN0642
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress α-L-Rhamnose monohydrate is a component of the plant cell wall pectic polysaccharides rhamnogalacturonan I and rhamnogalacturonan II. α-L-Rhamnose monohydrate is also a component of bacterial polysaccharides where it plays an important role in pathogenicity.

α-L-Rhamnose monohydrate is a component of the plant cell wall pectic polysaccharides rhamnogalacturonan I and rhamnogalacturonan II. α-L-Rhamnose monohydrate is also a component of bacterial polysaccharides where it plays an important role in pathogenicity.

Specifications

Chemical Name or Material α-L-Rhamnose monohydrate
CAS 6155-35-7
Color White
Purity Grade Notes Research
Molecular Formula C6H14O6
Quantity 100 mg
Solubility Information H2O : 100 mg/mL (548.94 mM; Need ultrasonic and warming)
SMILES O[C@@H]([C@@H](O[C@H]1O)C)[C@H]([C@H]1O)O.O
Molecular Weight (g/mol) 182.17
Formula Weight 182.17
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.