missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ L-Quisqualic acid

Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
Description
L-Quisqualic acid is a glutamate receptor agonist acting at AMPA receptors and metabotropic glutamate receptors positively linked to phosphoinositide hydrolysis. L-Quisqualic acid sensitizes neurons in hippocampus to depolarization by L-AP6 (the so called 'quis' effect).
Specifications
Specifications
| Quantity | 1 mg |
| Formula Weight | 189.13 |
| Purity | >99% |
| Molecular Weight (g/mol) | 189.13 |
| Chemical Name or Material | L-Quisqualic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction