missing translation for 'onlineSavingsMsg'
Learn More

L-Pyroglutamic acid, MedChemExpress

Product Code. 16451969
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451969 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16451969 Supplier Medchem Express Supplier No. HY76082
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress L-Pyroglutamic acid is the levo-isomer of Pyroglutamic acid. L-Pyroglutamic acid is the biologically active enantiomer in humans. Pyroglutamic acid is an intermediate in glutathione metabolism.

L-Pyroglutamic acid is the levo-isomer of Pyroglutamic acid. L-Pyroglutamic acid is the biologically active enantiomer in humans. Pyroglutamic acid is an intermediate in glutathione metabolism.

Specifications

Chemical Name or Material L-Pyroglutamic acid
CAS 98-79-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C5H7NO3
Solubility Information DMSO : 100 mg/mL (774.53 mM; Need ultrasonic) ∣H2O : 100 mg/mL (774.53 mM; Need ultrasonic)
SMILES O=C(O)[C@H](CC1)NC1=O
Molecular Weight (g/mol) 129.11
Quantity 1 g
Formula Weight 129.11
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.